Order Entry
Austria
ContactUsLinkComponent
Strontium standard for ion chromatography, (CRM), Fluka™
Strontium standard for ion chromatography, (CRM), Fluka™
  HONCE3045-100ML
 :  
Strontium standard for ion chromatography, (CRM), Fluka™
  HONCE3045-100ML
 :  

 

  • Concentration:
    1000 mg/L
  • Matrix:
    H₂O
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Element:
    Strontium
  • Symbol:
    Sr

 

 

Certified Reference Material (CRM)

  • Traceable to SRM from NIST
 : ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).