Order Entry
Austria
ContactUsLinkComponent
Iridium ICP and ICP-MS Standards, (CRMs), Fluka™
Iridium ICP and ICP-MS Standards, (CRMs), Fluka™
Catalog # HONCE1326-100ML
CAS Number:  
Iridium ICP and ICP-MS Standards, (CRMs), Fluka™
Catalog # HONCE1326-100ML
CAS Number:  

Specifications

  • Concentration:
    100 mg/L
  • Matrix:
    2% HCl
  • Pack type:
    Plastic bottle
  • Pk:
    100 ml
  • Element:
    Iridium
  • Symbol:
    Ir

Specifications

About this item

Certified Reference Materials (CRMs)

  • Traceable to SRM from NIST
Certifications: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).