Order Entry
Austria
ContactUsLinkComponent
Holmium ICP & ICP-MS standards, (CRMs), Fluka™
Holmium ICP & ICP-MS standards, (CRMs), Fluka™
Catalog # HONCE1392-100ML
CAS Number:  
Holmium ICP & ICP-MS standards, (CRMs), Fluka™
Catalog # HONCE1392-100ML
CAS Number:  

Specifications

  • Concentration:
    10 mg/L
  • Matrix:
    2% HNO₃
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Element:
    Holmium
  • Symbol:
    Ho

Specifications

About this item

Certified Reference Materials (CRMs)

  • Traceable to SRM from NIST
Certifications: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).