Order Entry
Austria
ContactUsLinkComponent
Cadmium ICP & ICP-MS standard, (CRMs), Fluka™
Cadmium ICP & ICP-MS standard, (CRMs), Fluka™
Catalog # HONCE1309-100ML
CAS Number:  
Cadmium ICP & ICP-MS standard, (CRMs), Fluka™
Catalog # HONCE1309-100ML
CAS Number:  

Specifications

  • Concentration:
    100 mg/L
  • Matrix:
    2% HNO₃
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Element:
    Cadmium
  • Symbol:
    Cd

Specifications

About this item

Certified Reference Material (CRM)

  • Traceable to SRM from NIST
Certifications: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).