Niobium ICP standards, (CRMs), Fluka™
Supplier: Honeywell Chemicals
Certificates
About this item
Certified Reference Materials (CRMs)
- Traceable to SRM from NIST
Certifications: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).
Frequently Bought Together
ICP multi-element standards, CertiPUR®, Supelco®
Supplier: Merck
VWR® ARISTAR®, Multi-Element Calibration Standard IV in dil. Nitric Acid for ICP
Supplier: VWR Chemicals
VWR® AVS TITRINORM pH Buffer Solutions in Sachets, 20 °C
Supplier: VWR Chemicals
Conductivity standards, KCl, referenced at 25 °C, for conductivity measurement
Supplier: Mettler - Toledo
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
Xylenol orange triturate 1% (w/w) in potassium nitrate Reag. Ph. Eur. 1096301
Supplier: VWR Chemicals
pH buffers, ready to use
Supplier: WTW
Buffers, concentrated solutions, Titrisol®, Supelco®
Supplier: Merck
VWR® ARISTAR®, Multi-Element Standards, Anionic for Ion Chromatography
Supplier: VWR Chemicals
Buffer solutions in sachets, Certipur®, Supelco®
Supplier: Merck
Reagents for alkalinity determination using Lovibond® MD photometers
Supplier: Lovibond
Conductivity standards, Potassium chloride, IUPAC
Supplier: Hach
VWR® AVS TITRINORM Buffer Solutions, CRMs, at 25 °C
Supplier: VWR Chemicals
pH Standard Solutions, IUPAC, Certified
Supplier: Hach