Order Entry
Austria
ContactUsLinkComponent
Boron ICP standards, (CRMs), Fluka™
Boron ICP standards, (CRMs), Fluka™
Catalog # HONCE1134-100ML
CAS Number:  
Boron ICP standards, (CRMs), Fluka™
Catalog # HONCE1134-100ML
CAS Number:  

Specifications

  • Concentration:
    10000 mg/L
  • Matrix:
    H₂O
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Element:
    Boron
  • Symbol:
    B

Specifications

About this item

Certified Reference Materials (CRMs)

  • Traceable to SRM from NIST
Certifications: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).