Order Entry
Austria
ContactUsLinkComponent
NexION dual detector solution
NexION dual detector solution
Catalog # HONCE2195-100ML
CAS Number:  
NexION dual detector solution
Catalog # HONCE2195-100ML
CAS Number:  

Specifications

  • Matrix:
    2% HNO₃
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Description:
    NexION Dual Detector Solution

Specifications

About this item

Certified Reference Material (CRM).

200 g/L each of Al, Ba, Ce, Co, Cu, In, Li, Mg, Mn, Ni, Pb, Tb, U, Zn in HNO₃ 2%