NexION KED mode detection limit solution
Catalog # HONCE2194-100ML
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Matrix:1% HCl
- Pack type:Plastic bottle
- Pk:100 mL
- Description:NexION KED Mode Detection Limit Solution
Specifications
About this item
Certified Reference Material (CRM).
10 g/L each of V, As, Se in HCl 1%