Order Entry
Austria
ContactUsLinkComponent
NexION KED setup solution
NexION KED setup solution
Catalog # HONCE2189-100ML
CAS Number:  
NexION KED setup solution
Catalog # HONCE2189-100ML
CAS Number:  

Specifications

  • Matrix:
    1% HCl
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Description:
    NexION KED Setup Solution

Specifications

About this item

Certified Reference Material (CRM).

10 g/L Co; 1 g/L Ce in HCl 1%