Order Entry
Austria
ContactUsLinkComponent
Elan 9000/6100 setup stability masscal solution Hg for ICP
Elan 9000/6100 setup stability masscal solution Hg for ICP
Catalog # HONCE2183-100ML
CAS Number:  
Elan 9000/6100 setup stability masscal solution Hg for ICP
Catalog # HONCE2183-100ML
CAS Number:  

Specifications

  • Matrix:
    1% HNO₃
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Description:
    Elan 9000/6100 Setup Stability Masscal Solution Hg

Specifications

About this item

Certified Reference Material (CRM).

10 g/L each of Ba, Ce, Cd, Cu, In, Pb, Mg, Rh, U in HNO₃ 1%