Order Entry
Austria
ContactUsLinkComponent
Initial calibration verification standard 2 Hg for ICP
Initial calibration verification standard 2 Hg for ICP
Catalog # HONCE2181-100ML
CAS Number:  
Initial calibration verification standard 2 Hg for ICP
Catalog # HONCE2181-100ML
CAS Number:  

Specifications

  • Matrix:
    2% HNO₃
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Description:
    Initial Calibration Verification Standard 2 Hg

Specifications

About this item

Certified Reference Material (CRM).

10 mg/L each of Sn, Ti in HNO₃ 2%