Order Entry
Austria
ContactUsLinkComponent
 
Alternate Metals II for AA and ICP-OES
Catalog # HONCE2170-100ML
CAS Number:  
Alternate Metals II for AA and ICP-OES
Catalog # HONCE2170-100ML
CAS Number:  

Specifications

  • Matrix:
    2% HNO₃
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Description:
    Alternate Metals II

Specifications

About this item

Certified Reference Material (CRM).

500 mg/L each of Ca, Na, 100 mg/L each of K, Mg in HNO₃ 2%