Order Entry
Austria
ContactUsLinkComponent
Trace Metals I for AA and ICP-OES
Trace Metals I for AA and ICP-OES
Catalog # HONCE2166-100ML
CAS Number:  
Trace Metals I for AA and ICP-OES
Catalog # HONCE2166-100ML
CAS Number:  

Specifications

  • Matrix:
    5% HNO₃
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Description:
    Trace Metals I

Specifications

About this item

Certified Reference Material (CRM).

500 mg/L Al, 250 mg/L V, 100 mg/L each of As, Be, Co, Cr, Cu, Fe, Mn, Ni, Pb, Zn, 25 mg/L each of Cd, Se, 10 mg/L Hg in HNO₃ 5%