Order Entry
Austria
ContactUsLinkComponent
USP elemental impurities solution 1
USP elemental impurities solution 1
Catalog # HONCE2162-100ML
CAS Number:  
USP elemental impurities solution 1
Catalog # HONCE2162-100ML
CAS Number:  

Specifications

  • Matrix:
    5% HNO₃
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Description:
    USP Elemental Impurities Solution 1

Specifications

About this item

Certified Reference Material (CRM).

2500 mg/kg each of Cu, Mn, 250 mg/kg each of Cr, Mo, Ni, V, 15 mg/kg each of As, Hg, 10 mg/kg Pb, 5 mg/kg Cd in HNO₃ 5%