Order Entry
Austria
ContactUsLinkComponent
Analytes B for ICP
Analytes B for ICP
Catalog # HONCE2156-100ML
CAS Number:  
Analytes B for ICP
Catalog # HONCE2156-100ML
CAS Number:  

Specifications

  • Matrix:
    2% HNO₃
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Description:
    Analytes B

Specifications

About this item

Certified Reference Material (CRM).

20 mg/L each of Co, Cr, Cu, Mn, Ni, V, 10 mg/L each of As, Cd, Se, Zn, 5 mg/L Ag in HNO₃ 2%