Order Entry
Austria
ContactUsLinkComponent
Instrument calibration standard 1 for ICP
Instrument calibration standard 1 for ICP
Catalog # HONCE2148-100ML
CAS Number:  
Instrument calibration standard 1 for ICP
Catalog # HONCE2148-100ML
CAS Number:  

Specifications

  • Matrix:
    5% HNO₃
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Description:
    Instrument Calibration Standard 1

Specifications

About this item

Certified Reference Material (CRM).

20 mg/L each of Ag, Al, As, Ba, Be, Cd, Co, Cr, Cu, Mn, Mo, Ni, Pb, Sb, Se, Th, Tl, U, V, Zn in HNO₃ 5%