Order Entry
Austria
ContactUsLinkComponent
Multi-element solution for ICP, (4 components)
Multi-element solution for ICP, (4 components)
Catalog # HONCE2131-100ML
CAS Number:  
Multi-element solution for ICP, (4 components)
Catalog # HONCE2131-100ML
CAS Number:  

Specifications

  • Matrix:
    5% HNO₃
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Description:
    Multi-Element Solution for ICP, (4 components)

Specifications

About this item

Certified Reference Material (CRM).

1000 mg/L each of Al, Ca, Fe, Mg in HNO₃ 5%