Order Entry
Austria
ContactUsLinkComponent
 
Low UV standard for ICP
Catalog # HONCE2125-100ML
CAS Number:  
Low UV standard for ICP
Catalog # HONCE2125-100ML
CAS Number:  

Specifications

  • Matrix:
    2% HNO₃
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Description:
    Low UV Standard

Specifications

About this item

Certified Reference Material (CRM).

10 mg/L each of Al, P, S in HNO₃ 2%