Order Entry
Austria
ContactUsLinkComponent
Multi-element solution 5 for ICP
Multi-element solution 5 for ICP
Catalog # HONCE2121-100ML
CAS Number:  
Multi-element solution 5 for ICP
Catalog # HONCE2121-100ML
CAS Number:  

Specifications

  • Matrix:
    2% HNO₃
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Description:
    Multi-Element Solution 5

Specifications

About this item

Certified Reference Material (CRM).

10 mg/L each of B, Ge, Mo, Nb, P, Re, S, Si, Ta, Ti, W, Zr in HNO₃ 2%