Order Entry
Austria
ContactUsLinkComponent
 
Instrument Calibration standard 3 for ICP and HGA elements
Catalog # HONCE2105-100ML
CAS Number:  
Instrument Calibration standard 3 for ICP and HGA elements
Catalog # HONCE2105-100ML
CAS Number:  

Specifications

  • Matrix:
    5% HNO₃
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Description:
    Instrument Calibration Standard 3

Specifications

About this item

Certified Reference Material (CRM).

2000 mg/L each of Al, Ba, 1000 mg/L Fe, 500 mg/L each of Co, V, 250 mg/L Cu, 50 mg/L Be in HNO₃ 5%