Order Entry
Austria
ContactUsLinkComponent
Instrument Calibration standard 1 for ICP and HGA elements
Instrument Calibration standard 1 for ICP and HGA elements
Catalog # HONCE2103-100ML
CAS Number:  
Instrument Calibration standard 1 for ICP and HGA elements
Catalog # HONCE2103-100ML
CAS Number:  

Specifications

  • Matrix:
    5% HNO₃
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Description:
    Instrument Calibration Standard 1

Specifications

About this item

Certified Reference Material (CRM).

5000 mg/L each of Ca, K, Mg, Na in HNO₃ 5%