Order Entry
Austria
ContactUsLinkComponent
Mixed calibration standard 4 for ICP
Mixed calibration standard 4 for ICP
Catalog # HONCE2097-100ML
CAS Number:  
Mixed calibration standard 4 for ICP
Catalog # HONCE2097-100ML
CAS Number:  

Specifications

  • Matrix:
    5% HNO₃
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Description:
    Mixed Calibration Standard 4

Specifications

About this item

Certified Reference Material (CRM).

1000 mg/L Ca, 400 mg/L K, 200 mg/L each of Al, Na, 20 mg/L each of Cr, Ni in HNO₃ 5%