Order Entry
Austria
ContactUsLinkComponent
Mixed calibration standard 2 for ICP
Mixed calibration standard 2 for ICP
Catalog # HONCE2095-100ML
CAS Number:  
Mixed calibration standard 2 for ICP
Catalog # HONCE2095-100ML
CAS Number:  

Specifications

  • Matrix:
    5% HNO₃
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Description:
    Mixed Calibration Standard 2

Specifications

About this item

Certified Reference Material (CRM).

10 g/L Fe, 100mg/L each of Ba, Co, Cu, V in HNO₃ 5%