Order Entry
Austria
ContactUsLinkComponent
Mixed calibration standard 1 for ICP
Mixed calibration standard 1 for ICP
Catalog # HONCE2094-100ML
CAS Number:  
Mixed calibration standard 1 for ICP
Catalog # HONCE2094-100ML
CAS Number:  

Specifications

  • Matrix:
    2% HNO₃
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Description:
    Mixed Calibration Standard 1

Specifications

About this item

Certified Reference Material (CRM).

500 mg/L Pb, 200 mg/L Se, 150 mg/L each of Cd, Zn, 100 mg/L Mn, 50 mg/L Be in HNO₃ 2%