Order Entry
Austria
ContactUsLinkComponent
Interference check solution B for ICP-MS systems
Interference check solution B for ICP-MS systems
Catalog # HONCE2005-100ML
CAS Number:  
Interference check solution B for ICP-MS systems
Catalog # HONCE2005-100ML
CAS Number:  

Specifications

  • Matrix:
    5% HNO₃
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Description:
    Interference Check Solution B

Specifications

About this item

Certified Reference Material (CRM).

20 mg/L each of Cr, Co, Cu, Mn, Ni, V, 10 mg/L each of As, Cd, Se, Zn, 5 mg/L Ag in HNO₃ 5%