Order Entry
Austria
ContactUsLinkComponent
Calibration mix majors for AA and ICP-OES
Calibration mix majors for AA and ICP-OES
Catalog # HONCE2022-100ML
CAS Number:  
Calibration mix majors for AA and ICP-OES
Catalog # HONCE2022-100ML
CAS Number:  

Specifications

  • Matrix:
    5% HNO₃
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Description:
    Calibration Mix Majors

Specifications

About this item

Certified Reference Material (CRM).

500 mg/L each of Ca, Fe, K, Mg, Na in HNO₃ 5%