Order Entry
Austria
ContactUsLinkComponent
Calibration mix 1 for AA and ICP-OES
Calibration mix 1 for AA and ICP-OES
Catalog # HONCE2020-100ML
CAS Number:  
Calibration mix 1 for AA and ICP-OES
Catalog # HONCE2020-100ML
CAS Number:  

Specifications

  • Matrix:
    2% HNO₃/0,5% HF
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Description:
    Calibration Mix 1

Specifications

About this item

Certified Reference Material (CRM).

100 mg/L each of Mo, Sb, Sn, Ti in HNO₃ 2%, HF 0.5%