Order Entry
Austria
ContactUsLinkComponent
ICP internal standard
ICP internal standard
Catalog # HONCE2019-100ML
CAS Number:  
ICP internal standard
Catalog # HONCE2019-100ML
CAS Number:  

Specifications

  • Matrix:
    5% HNO₃
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Description:
    ICP Internal Standard

Specifications

About this item

Certified Reference Material (CRM).

100 mg/L each of Bi, In, 6Li, Sc, Tb, Y in HNO₃ 5%