Order Entry
Austria
ContactUsLinkComponent
Internal standard mix for ICP-MS systems
Internal standard mix for ICP-MS systems
Catalog # HONCE2006-100ML
CAS Number:  
Internal standard mix for ICP-MS systems
Catalog # HONCE2006-100ML
CAS Number:  

Specifications

  • Matrix:
    10% HNO₃
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Description:
    Internal Standard Mix

Specifications

About this item

Certified Reference Material (CRM).

100 mg/L each of Bi, Ge, In, 6Li, Lu, Rh, Sc, Tb in HNO₃ 10%