Order Entry
Austria
ContactUsLinkComponent
Interference check solution A for ICP-MS systems
Interference check solution A for ICP-MS systems
Catalog # HONCE2004-100ML
CAS Number:  
Interference check solution A for ICP-MS systems
Catalog # HONCE2004-100ML
CAS Number:  

Specifications

  • Matrix:
    5% HNO₃
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Description:
    Interference Check Solution A

Specifications

About this item

Certified Reference Material (CRM).

20 g/L Cl-, 3000 mg/L Ca, 2500 mg/L each of Fe, Na; 2000 mg/L C, 1000 mg/L each of Al, Mg, P, K, S, 20 mg/L each of Mo, Ti in HNO₃ 5%