Trifluoroacetic acid ≥99.8% (by anhydrous basis), SPECTRONORM® for spectroscopy
: TFA
:76-05-1
:
:C₂HF₃O₂
:MFCD00004169
:114.02 g/mol
:–15 °C
:2699
:8,I
:Packaged under nitrogen.
:GHS07
:GHS05
:Ambient
- Environmentally Preferable:
- Assay (on anhydrous substance):min 99.8%
- Water:max 0.10%
- Colour number (APHA):max 10
- Evaproration residue:max 0.005%
- Transmittance (at 265 nm):min 10%
- Transmittance (at 305 nm):min 50%
- Transmittance (at 320 nm):min 80%
- Transmittance (at 325 nm):min 90%
- manufactured and filled under nitrogen:Yes
Frequently Bought Together