Handheld refractometer, Acidity meter for vinegar, PAL- Easy ACID 181 master kit
Catalog # 635-0833
Supplier: Atago
Specifications
- Description:Acidity meter for vinegar
- Pk:1
Specifications
About this item
This Master Kit is specifically for measuring vinegar and includes everything you need for easy, precise and accurate measurements.
- ATAGO's durable titan electrode will show the result that correlated with that of titaration
To measure acidity, sample needs to be dilulted.
Ordering information: Requires 2 x AAA alkaline batteries.
Delivery: The master kit comes complete with a scale, 1 beaker, and 1 measuring spoon.