ContactUsLinkComponent
Handheld refractometer for ethyl alcohol, PAL-Covid-19
Handheld refractometer for ethyl alcohol, PAL-Covid-19
Catalog # ATAG4553
Supplier:  Atago
Handheld refractometer for ethyl alcohol, PAL-Covid-19
Catalog # ATAG4553
Supplier:  Atago
Specifications
  • Description:
    Ethyl alcohol refractometer, PAL-Covid-19
  • Pk:
    1
Specifications
About this item
PAL-Covid19 will measure the concentration of alcohol percentage for hand made sanitizer.
  • Measure ethanol concentration easily
You can easily measure the alcohol concentration with PAL-Covid19 in 4 steps. Low alcohol concentration is ineffective for sterlization, but high alcohol conentration may cause rough skin.
Ordering information: Requires 2 AAA batteries.