Order Entry
Austria
ContactUsLinkComponent
 
Ammonium dihydrogen phosphate 100 g/L in nitric acid 1% matrix modifier solution, for atomic absorption spectroscopy, Fluka™
  HONCE4606-50ML
 :  
Ammonium dihydrogen phosphate 100 g/L in nitric acid 1% matrix modifier solution, for atomic absorption spectroscopy, Fluka™
  HONCE4606-50ML
 :  

 

  • Pack type:
    Plastic bottle
  • Pk:
    50 mL
  • Seq:
    2370

 

 

  • Matrix:
    HNO₃ 1%
  • Concentration:
    NH₄H₂PO₄ 100 g/L
  • Quality:
    ISO/IEC17025:2017(ANAB),ISO17034:2016(ANAB)
  • Form:
    Solution
  • Density:
    1.01g/cm3

 

 

 :
 :1.01 g/cm3
 :115.03 g/mol
 :Ambient
 :7722-76-1
 :NH₄H₂PO₄
 :MFCD00003396