Order Entry
Austria
ContactUsLinkComponent
Sera-Xtracta virus/pathogen kit
Sera-Xtracta virus/pathogen kit
Supplier:  Cytiva
Certificates
undefined
Sera-Xtracta virus/pathogen kit
Supplier:  Cytiva

Specifications

  • Method / Format:
    Magnetic beads
  • Prep size:
    Scalable
  • Refine target molecule:
    Viral RNA
    Bacterial RNA
    Viral DNA
    Bacterial DNA
  • Sample type:
    Swabs
    Blood
    Biofluids (serum, plasma, saliva)
  • Target molecule:
    Copurification of DNA & RNA
    DNA
    RNA
  • Description:
    Sera-Xtracta virus/pathogen kit

Specifications

Related Information

Frequently Bought Together