Disodium hydrogen phosphate
Supplier: Merck
Jump to:
About this item
Synonyms: sec-Sodium phosphate, di-Sodium hydrogen orthophosphate, di-Sodium hydrogen phosphate anhydrous, Sodium hydrogen phosphate, Disodium phosphate, Sodium phosphate dibasic
- CAS Number:7558-79-4
- Formula:Na₂HPO₄
- MDL Number:MFCD00003496
- Molecular Weight:141.96 g/mol
- :M-Clarity™ quality level = MQ100
2 Options Available Below
Product Details & Documents
Synonyms: sec-Sodium phosphate, di-Sodium hydrogen orthophosphate, di-Sodium hydrogen phosphate anhydrous, Sodium hydrogen phosphate, Disodium phosphate, Sodium phosphate dibasic
- CAS Number:7558-79-4
- Formula:Na₂HPO₄
- MDL Number:MFCD00003496
- Molecular Weight:141.96 g/mol
- :M-Clarity™ quality level = MQ100
Product Family Options
Product Information
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



