The IKA Matrix Orbital Delta family of thermoshakers, with mixing and heating, have numerous user-friendly features.
- Large, clear display with convenient menu structure allows for intuitive operation
- Speeds of up to 2000 min⁻¹ (1500 for the Δ F1.5 and Δ F2.0 models), and shaking diameter of 3 mm ensures the liquid mixes quickly and effectively
- Two different speed modes for gentle heating of samples
- Programmable ramps allow for automatic execution of mixing and heating tasks
- Smart speed adjustment according to insert, attachment and filling volume
- Aluminium die-cast housing ensures secure positioning, excellent stability and everyday robustness
- RS232 and USB interfaces
Whether it is blood samples, pharmaceutical agents, DNA/RNA samples or ELISA assays – samples with even the smallest volumes mix reliably and completely in all laboratory applications. No cross-contamination and an optimal mixing result.
IKA Matrix Orbital Δ FP mixes, heats and has a fixed universal holder for microtiter plates, a secure, quick lock for fast and easy change of inserts and attachments, as well as automatic attachment detection.
IKA Matrix Orbital Δ F2.0 mixes and heats with a fixed insert for 24×2,0 ml tubes.
IKA Matrix Orbital Δ F1.5 mixes and heats with a fixed insert for 24×1,5 ml tubes.
IKA Matrix Orbital Δ F0.5 mixes and heats with a fixed insert for 24×0,5 ml tubes.
- Catalog No:
- 444-1279
- 444-1280
- 444-1278
- 444-1281
- Model:
- IKA Matrix Orbital Delta F1.5
- IKA Matrix Orbital Delta FP
- IKA Matrix Orbital Delta F0.5
- IKA Matrix Orbital Delta F2.0
- Incubating accuracy:
- ±0,5 at 20...45
- ±1 at 20...45 °C
- ±0,5 at 20...45 °C
- ±0,5 at 20...45 °C
- Temperature control range:
- Ambient +4...100 °C
- Ambient +4...100 °C
- Ambient +4...100 °C
- Ambient +4...100 °C
- Heating speed:
- 11 °C/min
- 18 °C/min
- 15 °C/min
- 13 °C/min
- W×D×H:
- 217×158×351 mm
- 216×147×351 mm
- 216×149×351 mm
- 216×158×351 mm
- Weight:
- 9,2 kg
- 9,2 kg
- 9,2 kg
- 9,2 kg
- Orbit:
- 3 mm
- 3 mm
- 3 mm
- 3 mm
- Mixing speed range:
- 0 - 1500 min⁻¹
- 0 - 2000 min⁻¹
- 0 - 2000 min⁻¹
- 0 - 1500 min⁻¹