Bromothymol blue, ANALAR NORMAPUR® ACS
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:76-59-5
:
Formula:C₂₇H₂₈Br₂O₅S
MDL Number:MFCD00005872
:624.39 g/mol
Specification Test Results
- Clarity of solution:Passes test ACS
- Identification (UV/VIS) :Passes test
- Sensitivity:Passes test
- Visual transition interval:pH 5.8 - pH 7.6
- Loss on drying (110°C):Max. 3 %
- Absorption max. λ 1 max (buffer pH 5.8):430 - 435 nm
- Absorption max. λ 2 max (buffer pH 7.6):615 - 618 nm
- Sp.absorptiv.(λ1max;0.005g/l;pH5.8;dry):260 - 305
- Sp.absorptiv.(λ2max;0.005g/l;pH7.6;dry) :455 - 530
Specification Test Results
Related Information
Frequently Bought Together