Order Entry
Austria
ContactUsLinkComponent
Upright microscope, DM750
Upright microscope, DM750
Supplier:  LEICA MICROSYSTEMS
Certificates
undefined
Upright microscope, DM750
Supplier:  LEICA MICROSYSTEMS

Specifications

  • Catalog No:
  • 630-3283
  • 630-3280
  • 630-3281
  • 630-3282
  • 630-3277
  • 630-3284
  • 630-3278
  • 630-3279
  • Model:
  • DM750 package 7
  • DM750 package 4
  • DM750 package 5
  • DM750 package 6
  • DM750 package 1
  • DM750 package 8
  • DM750 package 2
  • DM750 package 3
  • Observation method:
  • Brightfield and phase contrast
  • Brightfield
  • Brightfield and phase contrast
  • Brightfield and phase contrast
  • Brightfield
  • Brightfield and phase contrast
  • Brightfield
  • Brightfield
  • Head:
  • 45° inclined
  • 45° inclined
  • 45° inclined
  • 45° inclined
  • 45° inclined
  • 45° inclined
  • 45° inclined
  • 45° inclined
  • Eyepieces:
  • WF 10×/20 mm
  • WF 10×/20 mm
  • WF 10×/20 mm
  • WF 10×/20 mm
  • WF 10×/20 mm
  • WF 10×/20 mm
  • WF 10×/20 mm
  • WF 10×/20 mm
  • Objectives:
  • Hi Plan 10× PH1, 20× PH1, 40× PH1, 100× PH3 (i)
  • Hi Plan 4×, 10×, 40×, 100× (i)
  • Hi Plan 10× PH1, 20× PH1, 40× PH1, 100× PH3 (i)
  • Hi Plan 10× PH1, 20× PH1, 40× PH1, 100× PH3 (i)
  • Hi Plan 4×, 10×, 40×, 100× (i)
  • Hi Plan 10× PH1, 20× PH1, 40× PH1, 100× PH3 (i)
  • Hi Plan 4×, 10×, 40×, 100× (i)
  • Hi Plan 4×, 10×, 40×, 100× (i)
  • Condenser:
  • Abbe condenser 0,9 dry/1,25 oil with slot for phase and darkfield sliders
  • Abbe condenser 0,9 dry/1,25 oil with slot for phase and darkfield sliders
  • Abbe condenser 0,9 dry/1,25 oil with slot for phase and darkfield sliders
  • Abbe condenser 0,9 dry/1,25 oil with slot for phase and darkfield sliders
  • Abbe condenser 0,9 dry/1,25 oil with slot for phase and darkfield sliders
  • Abbe condenser 0,9 dry/1,25 oil with slot for phase and darkfield sliders
  • Abbe condenser 0,9 dry/1,25 oil with slot for phase and darkfield sliders
  • Abbe condenser 0,9 dry/1,25 oil with slot for phase and darkfield sliders
  • Stand:
  • RH
  • RH Koehler
  • RH
  • RH Koehler
  • RH
  • RH Koehler
  • RH Koehler
  • RH

Specifications

Objectives

Product Information

Frequently Bought Together