ContactUsLinkComponent
X-ray and metal detectable powder funnels
X-ray and metal detectable powder funnels
  SAMPM1051B-200
X-ray and metal detectable powder funnels
  SAMPM1051B-200
  • Ø:
    200 mm
  • Description:
    X-ray and metal detectable powder funnels, blue
  • Pk:
    4
X-ray and metal detectable powder funnels are ideal for both laboratory and production work. The bright blue colour makes the bottles easy to spot if they are left in the product.
  • Robust PP construction
  • Non sterile
  • PP gives excellent chemical resistance to many products
  • Full batch traceability
To ensure the cleanliness of the funnels, they are manufactured and packed in a cleanroom. The robust nature of these funnels means that they can give many years of service or for more demanding applications. They can be used as a single use disposable product straight from the bag.
Ideal for powders, creams, liquids and suspensions.
 : These PP funnels conform to all the latest standards including FDA 177.1520, EU 10/2011, EC 1935/2004 and are BSE/TSE free. Batch certificates are available.
 : Supplied individually bagged.