ContactUsLinkComponent
Avantor® Hichrom HI-SE54 HT, GC Columns
Avantor® Hichrom HI-SE54 HT, GC Columns
Supplier:  Avantor
Certificates
undefined
Avantor® Hichrom HI-SE54 HT, GC Columns
Supplier:  Avantor
Specifications
  • Catalog No:
  • HI91-32-025-30
  • HI91-53-010-10
  • HI91-10-010-10
  • HI91-10-010-5
  • HI91-53-015-15
  • HI91-53-015-30
  • HI91-53-010-15
  • HI91-53-010-30
  • HI91-53-025-30
  • HI91-53-017-30
  • HI91-53-025-15
  • HI91-25-010-15
  • HI91-25-010-30
  • HI91-32-010-10G2
  • HI91-25-015-15
  • HI91-25-025-30
  • HI91-32-010-10
  • HI91-25-015-30
  • HI91-25-025-15
  • HI91-32-015-15
  • HI91-32-015-30
  • HI91-32-010-15
  • HI91-32-010-30
  • HI91-32-025-15
  • Phase name:
  • HI-SE54 HT
  • HI-SE54 HT
  • HI-SE54 HT
  • HI-SE54 HT
  • HI-SE54 HT
  • HI-SE54 HT
  • HI-SE54 HT
  • HI-SE54 HT
  • HI-SE54 HT
  • HI-SE54 HT
  • HI-SE54 HT
  • HI-SE54 HT
  • HI-SE54 HT
  • HI-SE54 HT
  • HI-SE54 HT
  • HI-SE54 HT
  • HI-SE54 HT
  • HI-SE54 HT
  • HI-SE54 HT
  • HI-SE54 HT
  • HI-SE54 HT
  • HI-SE54 HT
  • HI-SE54 HT
  • HI-SE54 HT
  • Phase composition:
  • 5% Phenyl, 1% Vinyl, 94% Methyl Polysiloxane - high temperature
  • 5% Phenyl, 1% Vinyl, 94% Methyl Polysiloxane - high temperature
  • 5% Phenyl, 1% Vinyl, 94% Methyl Polysiloxane - high temperature
  • 5% Phenyl, 1% Vinyl, 94% Methyl Polysiloxane - high temperature
  • 5% Phenyl, 1% Vinyl, 94% Methyl Polysiloxane - high temperature
  • 5% Phenyl, 1% Vinyl, 94% Methyl Polysiloxane - high temperature
  • 5% Phenyl, 1% Vinyl, 94% Methyl Polysiloxane - high temperature
  • 5% Phenyl, 1% Vinyl, 94% Methyl Polysiloxane - high temperature
  • 5% Phenyl, 1% Vinyl, 94% Methyl Polysiloxane - high temperature
  • 5% Phenyl, 1% Vinyl, 94% Methyl Polysiloxane - high temperature
  • 5% Phenyl, 1% Vinyl, 94% Methyl Polysiloxane - high temperature
  • 5% Phenyl, 1% Vinyl, 94% Methyl Polysiloxane - high temperature
  • 5% Phenyl, 1% Vinyl, 94% Methyl Polysiloxane - high temperature
  • 5% Phenyl, 1% Vinyl, 94% Methyl Polysiloxane - high temperature
  • 5% Phenyl, 1% Vinyl, 94% Methyl Polysiloxane - high temperature
  • 5% Phenyl, 1% Vinyl, 94% Methyl Polysiloxane - high temperature
  • 5% Phenyl, 1% Vinyl, 94% Methyl Polysiloxane - high temperature
  • 5% Phenyl, 1% Vinyl, 94% Methyl Polysiloxane - high temperature
  • 5% Phenyl, 1% Vinyl, 94% Methyl Polysiloxane - high temperature
  • 5% Phenyl, 1% Vinyl, 94% Methyl Polysiloxane - high temperature
  • 5% Phenyl, 1% Vinyl, 94% Methyl Polysiloxane - high temperature
  • 5% Phenyl, 1% Vinyl, 94% Methyl Polysiloxane - high temperature
  • 5% Phenyl, 1% Vinyl, 94% Methyl Polysiloxane - high temperature
  • 5% Phenyl, 1% Vinyl, 94% Methyl Polysiloxane - high temperature
  • Polarity:
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Low-polarity stationary phase
  • Crossbond:
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Temperature limit:
  • 400 °C
  • 400 °C
  • 400 °C
  • 400 °C
  • 400 °C
  • 400 °C
  • 400 °C
  • 400 °C
  • 400 °C
  • 400 °C
  • 400 °C
  • 400 °C
  • 400 °C
  • 400 °C
  • 400 °C
  • 400 °C
  • 400 °C
  • 400 °C
  • 400 °C
  • 400 °C
  • 400 °C
  • 400 °C
  • 400 °C
  • 400 °C
  • USP code:
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41
  • G27, G36, G41