Avantor® Hichrom HI-17 HT, GC Columns
Supplier: Avantor
Certificates
About this item
High Temperature general purpose column. Ideal for confrmation analysis, ideal as high polarity dimension in GCxGC-HT confgurations.
Max temperature limits may change depending on the film thickness.
Specifications
- Catalog No:
- HI90-10-010-5
- HI90-32-025-30
- HI90-25-015-15
- HI90-53-010-10
- HI90-10-010-10
- HI90-53-015-15
- HI90-25-025-30
- HI90-32-010-10
- HI90-53-015-30
- HI90-53-010-15
- HI90-25-015-30
- HI90-53-010-30
- HI90-25-025-15
- HI90-53-025-30
- HI90-32-015-15
- HI90-32-015-30
- HI90-53-017-30
- HI90-32-010-15
- HI90-53-025-15
- HI90-32-010-30
- HI90-32-025-15
- HI90-25-010-15
- HI90-25-010-30
- Phase name:
- HI-17 HT
- HI-17 HT
- HI-17 HT
- HI-17 HT
- HI-17 HT
- HI-17 HT
- HI-17 HT
- HI-17 HT
- HI-17 HT
- HI-17 HT
- HI-17 HT
- HI-17 HT
- HI-17 HT
- HI-17 HT
- HI-17 HT
- HI-17 HT
- HI-17 HT
- HI-17 HT
- HI-17 HT
- HI-17 HT
- HI-17 HT
- HI-17 HT
- HI-17 HT
- Phase composition:
- 50% Phenyl, 50% Methyl Polysiloxane - high temperature
- 50% Phenyl, 50% Methyl Polysiloxane - high temperature
- 50% Phenyl, 50% Methyl Polysiloxane - high temperature
- 50% Phenyl, 50% Methyl Polysiloxane - high temperature
- 50% Phenyl, 50% Methyl Polysiloxane - high temperature
- 50% Phenyl, 50% Methyl Polysiloxane - high temperature
- 50% Phenyl, 50% Methyl Polysiloxane - high temperature
- 50% Phenyl, 50% Methyl Polysiloxane - high temperature
- 50% Phenyl, 50% Methyl Polysiloxane - high temperature
- 50% Phenyl, 50% Methyl Polysiloxane - high temperature
- 50% Phenyl, 50% Methyl Polysiloxane - high temperature
- 50% Phenyl, 50% Methyl Polysiloxane - high temperature
- 50% Phenyl, 50% Methyl Polysiloxane - high temperature
- 50% Phenyl, 50% Methyl Polysiloxane - high temperature
- 50% Phenyl, 50% Methyl Polysiloxane - high temperature
- 50% Phenyl, 50% Methyl Polysiloxane - high temperature
- 50% Phenyl, 50% Methyl Polysiloxane - high temperature
- 50% Phenyl, 50% Methyl Polysiloxane - high temperature
- 50% Phenyl, 50% Methyl Polysiloxane - high temperature
- 50% Phenyl, 50% Methyl Polysiloxane - high temperature
- 50% Phenyl, 50% Methyl Polysiloxane - high temperature
- 50% Phenyl, 50% Methyl Polysiloxane - high temperature
- 50% Phenyl, 50% Methyl Polysiloxane - high temperature
- Polarity:
- Mid-to-high polarity stationary phase
- Mid-to-high polarity stationary phase
- Mid-to-high polarity stationary phase
- Mid-to-high polarity stationary phase
- Mid-to-high polarity stationary phase
- Mid-to-high polarity stationary phase
- Mid-to-high polarity stationary phase
- Mid-to-high polarity stationary phase
- Mid-to-high polarity stationary phase
- Mid-to-high polarity stationary phase
- Mid-to-high polarity stationary phase
- Mid-to-high polarity stationary phase
- Mid-to-high polarity stationary phase
- Mid-to-high polarity stationary phase
- Mid-to-high polarity stationary phase
- Mid-to-high polarity stationary phase
- Mid-to-high polarity stationary phase
- Mid-to-high polarity stationary phase
- Mid-to-high polarity stationary phase
- Mid-to-high polarity stationary phase
- Mid-to-high polarity stationary phase
- Mid-to-high polarity stationary phase
- Mid-to-high polarity stationary phase
- Crossbond:
- Stabilised
- Stabilised
- Stabilised
- Stabilised
- Stabilised
- Stabilised
- Stabilised
- Stabilised
- Stabilised
- Stabilised
- Stabilised
- Stabilised
- Stabilised
- Stabilised
- Stabilised
- Stabilised
- Stabilised
- Stabilised
- Stabilised
- Stabilised
- Stabilised
- Stabilised
- Stabilised
- Temperature limit:
- 370 °C
- 370 °C
- 370 °C
- 370 °C
- 370 °C
- 370 °C
- 370 °C
- 370 °C
- 370 °C
- 370 °C
- 370 °C
- 370 °C
- 370 °C
- 370 °C
- 370 °C
- 370 °C
- 370 °C
- 370 °C
- 370 °C
- 370 °C
- 370 °C
- 370 °C
- 370 °C
- USP code:
- G3, G17
- G3, G17
- G3, G17
- G3, G17
- G3, G17
- G3, G17
- G3, G17
- G3, G17
- G3, G17
- G3, G17
- G3, G17
- G3, G17
- G3, G17
- G3, G17
- G3, G17
- G3, G17
- G3, G17
- G3, G17
- G3, G17
- G3, G17
- G3, G17
- G3, G17
- G3, G17