Avantor® Hichrom HI-TCEP, GC Columns
: Avantor
Application specifc columns ideal for aromatics and oxygenates in gasoline.
Max temperature limits may change depending on the film thickness.
- Catalog No:
- HI61-25-040-30
- HI61-32-051-60
- HI61-25-040-60
- HI61-25-044-60
- Phase name:
- HI-TCEP
- HI-TCEP
- HI-TCEP
- HI-TCEP
- Phase composition:
- 1,2,3-tris-(2-cyanoethoxy)propane
- 1,2,3-tris-(2-cyanoethoxy)propane
- 1,2,3-tris-(2-cyanoethoxy)propane
- 1,2,3-tris-(2-cyanoethoxy)propane
- Polarity:
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- High-polarity stationary phase
- Crossbond:
- Stabilised
- Stabilised
- Stabilised
- Stabilised
- Temperature limit:
- 150 °C
- 150 °C
- 150 °C
- 150 °C
Frequently Bought Together