ContactUsLinkComponent
Avantor® Hichrom HI-65 HT, GC Columns
Avantor® Hichrom HI-65 HT, GC Columns
Supplier:  Avantor
Certificates
undefined
Avantor® Hichrom HI-65 HT, GC Columns
Supplier:  Avantor
Specifications
  • Catalog No:
  • HI47-32-010-15
  • HI47-32-010-30
  • HI47-25-010-15
  • HI47-25-010-30
  • HIGCHI47-25-010-25
  • HI47-53-010-30
  • Phase name:
  • HI-65 HT
  • HI-65 HT
  • HI-65 HT
  • HI-65 HT
  • HI-65 HT
  • Phase composition:
  • 65% Phenyl, 35% Methyl Polysiloxane - high temperature
  • 65% Phenyl, 35% Methyl Polysiloxane - high temperature
  • 65% Phenyl, 35% Methyl Polysiloxane - high temperature
  • 65% Phenyl, 35% Methyl Polysiloxane - high temperature
  • 65% Phenyl, 35% Methyl Polysiloxane - high temperature
  • Polarity:
  • Polar stationary phase
  • Polar stationary phase
  • Polar stationary phase
  • Polar stationary phase
  • Polar stationary phase
  • Crossbond:
  • Yes
  • Yes
  • Yes
  • Yes
  • Yes
  • Temperature limit:
  • 370 °C
  • 370 °C
  • 370 °C
  • 370 °C
  • 370 °C