ContactUsLinkComponent
Avantor® Hichrom HI-WAX Spirit, GC Columns
Avantor® Hichrom HI-WAX Spirit, GC Columns
Supplier:  Avantor
Certificates
undefined
Avantor® Hichrom HI-WAX Spirit, GC Columns
Supplier:  Avantor
Specifications
  • Catalog No:
  • HIGCHI15-25-020-50
  • HI15-32-050-50
  • HIGCHI15-25-020-25
  • Phase name:
  • HI-WAX Spirit
  • HI-WAX Spirit
  • HI-WAX Spirit
  • Phase composition:
  • Polyethylene Glycol (PEG) designed for Spirits Analysis
  • Polyethylene Glycol (PEG) designed for Spirits Analysis
  • Polyethylene Glycol (PEG) designed for Spirits Analysis
  • Polarity:
  • Polar stationary phase
  • Polar stationary phase
  • Polar stationary phase
  • Crossbond:
  • Yes
  • Yes
  • Yes
  • USP code:
  • G14, G15, G16, G20, G39
  • G14, G15, G16, G20, G39
  • G14, G15, G16, G20, G39