Avantor® Hichrom HI-VOC 1, GC Columns
Supplier: Avantor
Certificates
About this item
Application specifc columns for volatile organic compounds (VOCs), solvents and purgeable compounds. Due to the high max. temperature, they are ideal for two-parallel columns confg. in the same oven. A good alternative to the HI-624 giving different selectivity. It is a more robust than the HI-624 and can operate at higher temperatures.
Max temperature limits may change depending on the film thickness.
Specifications
- Catalog No:
- HI64-53-300-75
- HI64-32-200-30
- HI64-32-200-60
- HI64-53-500-30
- HI64-53-300-30
- HI64-53-300-60
- HI64-32-200-105
- HI64-32-300-30
- HI64-25-100-30
- HI64-32-300-60
- HI64-32-200-75
- HI64-53-500-60
- HI64-32-300-105
- HI64-53-500-75
- HI64-25-150-60
- HI64-32-500-30
- HI64-25-200-30
- HI64-25-100-60
- HI64-32-300-75
- HI64-25-150-30
- HI64-32-500-105
- HI64-25-200-60
- HI64-53-200-75
- HI64-25-300-30
- HI64-53-100-30
- HI64-25-300-60
- HI64-53-100-60
- HI64-32-500-60
- HI64-32-500-75
- HI64-32-100-60
- HI64-53-150-60
- HI64-32-100-75
- HI64-53-150-75
- HI64-32-100-105
- HI64-53-100-75
- HI64-32-100-30
- HI64-53-150-30
- HI64-32-150-60
- HI64-32-150-75
- HI64-32-150-105
- HI64-53-200-30
- HI64-32-150-30
- HI64-53-200-60
- Phase name:
- HI-VOC 1
- HI-VOC 1
- HI-VOC 1
- HI-VOC 1
- HI-VOC 1
- HI-VOC 1
- HI-VOC 1
- HI-VOC 1
- HI-VOC 1
- HI-VOC 1
- HI-VOC 1
- HI-VOC 1
- HI-VOC 1
- HI-VOC 1
- HI-VOC 1
- HI-VOC 1
- HI-VOC 1
- HI-VOC 1
- HI-VOC 1
- HI-VOC 1
- HI-VOC 1
- HI-VOC 1
- HI-VOC 1
- HI-VOC 1
- HI-VOC 1
- HI-VOC 1
- HI-VOC 1
- HI-VOC 1
- HI-VOC 1
- HI-VOC 1
- HI-VOC 1
- HI-VOC 1
- HI-VOC 1
- HI-VOC 1
- HI-VOC 1
- HI-VOC 1
- HI-VOC 1
- HI-VOC 1
- HI-VOC 1
- HI-VOC 1
- HI-VOC 1
- HI-VOC 1
- HI-VOC 1
- Phase composition:
- Proprietary Phase developed for Volatile Organic Compounds
- Proprietary Phase developed for Volatile Organic Compounds
- Proprietary Phase developed for Volatile Organic Compounds
- Proprietary Phase developed for Volatile Organic Compounds
- Proprietary Phase developed for Volatile Organic Compounds
- Proprietary Phase developed for Volatile Organic Compounds
- Proprietary Phase developed for Volatile Organic Compounds
- Proprietary Phase developed for Volatile Organic Compounds
- Proprietary Phase developed for Volatile Organic Compounds
- Proprietary Phase developed for Volatile Organic Compounds
- Proprietary Phase developed for Volatile Organic Compounds
- Proprietary Phase developed for Volatile Organic Compounds
- Proprietary Phase developed for Volatile Organic Compounds
- Proprietary Phase developed for Volatile Organic Compounds
- Proprietary Phase developed for Volatile Organic Compounds
- Proprietary Phase developed for Volatile Organic Compounds
- Proprietary Phase developed for Volatile Organic Compounds
- Proprietary Phase developed for Volatile Organic Compounds
- Proprietary Phase developed for Volatile Organic Compounds
- Proprietary Phase developed for Volatile Organic Compounds
- Proprietary Phase developed for Volatile Organic Compounds
- Proprietary Phase developed for Volatile Organic Compounds
- Proprietary Phase developed for Volatile Organic Compounds
- Proprietary Phase developed for Volatile Organic Compounds
- Proprietary Phase developed for Volatile Organic Compounds
- Proprietary Phase developed for Volatile Organic Compounds
- Proprietary Phase developed for Volatile Organic Compounds
- Proprietary Phase developed for Volatile Organic Compounds
- Proprietary Phase developed for Volatile Organic Compounds
- Proprietary Phase developed for Volatile Organic Compounds
- Proprietary Phase developed for Volatile Organic Compounds
- Proprietary Phase developed for Volatile Organic Compounds
- Proprietary Phase developed for Volatile Organic Compounds
- Proprietary Phase developed for Volatile Organic Compounds
- Proprietary Phase developed for Volatile Organic Compounds
- Proprietary Phase developed for Volatile Organic Compounds
- Proprietary Phase developed for Volatile Organic Compounds
- Proprietary Phase developed for Volatile Organic Compounds
- Proprietary Phase developed for Volatile Organic Compounds
- Proprietary Phase developed for Volatile Organic Compounds
- Proprietary Phase developed for Volatile Organic Compounds
- Proprietary Phase developed for Volatile Organic Compounds
- Proprietary Phase developed for Volatile Organic Compounds
- Crossbond:
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes