ContactUsLinkComponent
Avantor® Hichrom HI-LAP, GC Columns
Avantor® Hichrom HI-LAP, GC Columns
Supplier:  Avantor
Certificates
undefined
Avantor® Hichrom HI-LAP, GC Columns
Supplier:  Avantor
Specifications
  • Catalog No:
  • HI46-32-010-25
  • HI46-25-010-25
  • Phase name:
  • HI-LAP
  • HI-LAP
  • Phase composition:
  • Proprietary Phase developed for Lipid Analysis Phase, Saturated and Unsaturated Triglycerides, Sterols and Lipid analysis
  • Proprietary Phase developed for Lipid Analysis Phase, Saturated and Unsaturated Triglycerides, Sterols and Lipid analysis
  • Crossbond:
  • Yes
  • Yes
  • Temperature limit:
  • 370 °C
  • 370 °C