Orders LinkContactUsLinkComponent
Avantor® Hichrom HI-BASIC, GC Columns
Avantor® Hichrom HI-BASIC, GC Columns
Supplier:   Avantor
Avantor® Hichrom HI-BASIC, GC Columns
Supplier:  Avantor
Certificates
Avantor® Hichrom HI-BASIC, GC Columns
4 Options Available Below
Product Details & Documents
Application specific column for basic compouns (e.g. amines).
Max temperature limits may change depending on the film thickness.
Specifications for Product Family
Specifications
  • Catalog No:
  • HI38-25-015-25
  • HI38-32-025-25
  • HI38-32-025-30
  • HIGCHI38-32-025-60
  • Phase name:
  • HI-BASIC
  • HI-BASIC
  • HI-BASIC
  • HI-BASIC
  • Phase composition:
  • Proprietary Phase developed for aliphatic and aromatics Amines
  • Proprietary Phase developed for aliphatic and aromatics Amines
  • Proprietary Phase developed for aliphatic and aromatics Amines
  • Proprietary Phase developed for aliphatic and aromatics Amines
  • Crossbond:
  • Stabilised
  • Stabilised
  • Stabilised
  • Stabilised
Specifications
Product Family Options
Product Information
  • Column Format
    Film thickness
    Ø int.
    Length
    Availability
    Price
  • GC Column
    Capillary
    0,15 µm
    0,25 mm
    25 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 731,00 €
    731,00 €
    1 * 1 ST
       731,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    HI38-25-015-25
    Phase name
    HI-BASIC
    Phase composition
    Proprietary Phase developed for aliphatic and aromatics Amines
    Crossbond
    Stabilised
  • GC Column
    Capillary
    0,25 µm
    0,32 mm
    30 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 731,00 €
    731,00 €
    1 * 1 ST
       731,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    HI38-32-025-25
    Phase name
    HI-BASIC
    Phase composition
    Proprietary Phase developed for aliphatic and aromatics Amines
    Crossbond
    Stabilised
  • GC Column
    Capillary
    0,25 µm
    0,32 mm
    30 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 740,00 €
    740,00 €
    1 * 1 ST
       740,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    HI38-32-025-30
    Phase name
    HI-BASIC
    Phase composition
    Proprietary Phase developed for aliphatic and aromatics Amines
    Crossbond
    Stabilised
  • GC Column
    Capillary
    0,25 µm
    0,32 mm
    60 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 530,00 €
    1 530,00 €
    1 * 1 ST
       1 530,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    HIGCHI38-32-025-60
    Phase name
    HI-BASIC
    Phase composition
    Proprietary Phase developed for aliphatic and aromatics Amines
    Crossbond
    Stabilised
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Linear 2-ways Glass Press-Fit Connectors for fused silica GC columns, O-Ø 0,25 - 0,80 mm
    Linear 2-ways Glass Press-Fit Connectors for fused silica GC columns, O-Ø 0,25 - 0,80 mm
    HI0-10-PF-C
    Log in for availability
    1 * 10 ST - 117,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Linear 2-ways Glass Press-Fit Connectors for fused silica GC columns, O-Ø 0,25 - 0,80 mm
    Linear 2-ways Glass Press-Fit Connectors for fused silica GC columns, O-Ø 0,25 - 0,80 mm
    HI0-100-PF-C
    Log in for availability
    1 * 100 ST - 849,00 €
Product Information
  • Catalog #
    Availability
    Price
  • 3-ways Glass Press-Fit Connectors for fused silica GC columns, O-Ø 0,25 - 0,80 mm
    3-ways Glass Press-Fit Connectors for fused silica GC columns, O-Ø 0,25 - 0,80 mm
    HI0-10-PFY-C
    Log in for availability
    1 * 10 ST - 1 020,00 €
Product Information
  • Catalog #
    Availability
    Price
  • 3-ways Glass Press-Fit Connectors for fused silica GC columns, O-Ø 0,25 - 0,80 mm
    3-ways Glass Press-Fit Connectors for fused silica GC columns, O-Ø 0,25 - 0,80 mm
    HI0-1-PFY-C
    Log in for availability
    1 * 1 ST - 105,00 €
Product Information
  • Catalog #
    Availability
    Price
  • 3-ways Glass Press-Fit Connectors for fused silica GC columns, O-Ø 0,25 - 0,80 mm
    3-ways Glass Press-Fit Connectors for fused silica GC columns, O-Ø 0,25 - 0,80 mm
    HI0-2-PFY-C
    Log in for availability
    1 * 2 ST - 210,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Linear 2-ways Glass Press-Fit Connectors for fused silica GC columns, O-Ø 0,25 - 0,80 mm
    Linear 2-ways Glass Press-Fit Connectors for fused silica GC columns, O-Ø 0,25 - 0,80 mm
    HI0-25-PF-C
    Log in for availability
    1 * 25 ST - 226,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Linear 2-ways Glass Press-Fit Connectors for fused silica GC columns, O-Ø 0,25 - 0,80 mm
    Linear 2-ways Glass Press-Fit Connectors for fused silica GC columns, O-Ø 0,25 - 0,80 mm
    HI0-50-PF-C
    Log in for availability
    1 * 50 ST - 435,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Linear 2-ways Glass Press-Fit Connectors for fused silica GC columns, O-Ø 0,25 - 0,80 mm
    Linear 2-ways Glass Press-Fit Connectors for fused silica GC columns, O-Ø 0,25 - 0,80 mm
    HI0-5-PF-C
    Log in for availability
    1 * 5 ST - 78,00 €
Product Information
  • Catalog #
    Availability
    Price
  • 3-ways Glass Press-Fit Connectors for fused silica GC columns, O-Ø 0,25 - 0,80 mm
    3-ways Glass Press-Fit Connectors for fused silica GC columns, O-Ø 0,25 - 0,80 mm
    HI0-5-PFY-C
    Log in for availability
    1 * 5 ST - 517,00 €