ContactUsLinkComponent
Avantor® Hichrom HI-ALC 2, GC Columns
Avantor® Hichrom HI-ALC 2, GC Columns
Supplier:  Avantor
Certificates
undefined
Avantor® Hichrom HI-ALC 2, GC Columns
Supplier:  Avantor
Specifications
  • Catalog No:
  • HI37-53-100-30
  • HI37-32-060-30
  • HI37-32-100-30
  • Phase name:
  • HI-ALC 2
  • HI-ALC 2
  • HI-ALC 2
  • Phase composition:
  • Proprietary Phase developed for blood alcohol analysis
  • Proprietary Phase developed for blood alcohol analysis
  • Proprietary Phase developed for blood alcohol analysis
  • Crossbond:
  • Yes
  • Yes
  • Yes
  • Temperature limit:
  • 280 °C
  • 280 °C
  • 280 °C