Avantor® Hichrom HI-WAX HT, GC Columns
Supplier: Avantor
Certificates
About this item
High Temperature unique PEG phase. Extend the temperature limits of your FAST-GC and GCxGC methods while using a polar WAX phase.
Max temperature limits may change depending on the film thickness.
Specifications
- Catalog No:
- HI13-10-010-10
- HI13-53-010-10
- HI13-53-010-15
- HI13-10-010-5
- HI13-32-025-15
- HI13-32-025-30
- HI13-53-015-30
- HI13-53-017-30
- HI13-53-010-30
- HI13-53-015-15
- HI13-25-010-15
- HI13-53-025-15
- HI13-53-025-30
- HI13-25-015-15
- HI13-25-015-30
- HI13-25-010-30
- HI13-32-010-10
- HI13-32-010-15
- HI13-25-025-15
- HI13-25-025-30
- HI13-32-015-30
- HI13-32-010-30
- HIGCHI13-32-100-30
- HI13-32-015-15
- Phase name:
- HI-WAX HT
- HI-WAX HT
- HI-WAX HT
- HI-WAX HT
- HI-WAX HT
- HI-WAX HT
- HI-WAX HT
- HI-WAX HT
- HI-WAX HT
- HI-WAX HT
- HI-WAX HT
- HI-WAX HT
- HI-WAX HT
- HI-WAX HT
- HI-WAX HT
- HI-WAX HT
- HI-WAX HT
- HI-WAX HT
- HI-WAX HT
- HI-WAX HT
- HI-WAX HT
- HI-WAX HT
- HI-WAX HT
- HI-WAX HT
- Phase composition:
- Polyethylene Glycol (PEG) - high temperature
- Polyethylene Glycol (PEG) - high temperature
- Polyethylene Glycol (PEG) - high temperature
- Polyethylene Glycol (PEG) - high temperature
- Polyethylene Glycol (PEG) - high temperature
- Polyethylene Glycol (PEG) - high temperature
- Polyethylene Glycol (PEG) - high temperature
- Polyethylene Glycol (PEG) - high temperature
- Polyethylene Glycol (PEG) - high temperature
- Polyethylene Glycol (PEG) - high temperature
- Polyethylene Glycol (PEG) - high temperature
- Polyethylene Glycol (PEG) - high temperature
- Polyethylene Glycol (PEG) - high temperature
- Polyethylene Glycol (PEG) - high temperature
- Polyethylene Glycol (PEG) - high temperature
- Polyethylene Glycol (PEG) - high temperature
- Polyethylene Glycol (PEG) - high temperature
- Polyethylene Glycol (PEG) - high temperature
- Polyethylene Glycol (PEG) - high temperature
- Polyethylene Glycol (PEG) - high temperature
- Polyethylene Glycol (PEG) - high temperature
- Polyethylene Glycol (PEG) - high temperature
- Polyethylene Glycol (PEG) - high temperature
- Polyethylene Glycol (PEG) - high temperature
- Polarity:
- Polar stationary phase
- Polar stationary phase
- Polar stationary phase
- Polar stationary phase
- Polar stationary phase
- Polar stationary phase
- Polar stationary phase
- Polar stationary phase
- Polar stationary phase
- Polar stationary phase
- Polar stationary phase
- Polar stationary phase
- Polar stationary phase
- Polar stationary phase
- Polar stationary phase
- Polar stationary phase
- Polar stationary phase
- Polar stationary phase
- Polar stationary phase
- Polar stationary phase
- Polar stationary phase
- Polar stationary phase
- Polar stationary phase
- Polar stationary phase
- Crossbond:
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Temperature limit:
- 300 °C
- 300 °C
- 300 °C
- 300 °C
- 300 °C
- 300 °C
- 300 °C
- 300 °C
- 300 °C
- 300 °C
- 300 °C
- 300 °C
- 300 °C
- 300 °C
- 300 °C
- 300 °C
- 300 °C
- 300 °C
- 300 °C
- 300 °C
- 300 °C
- 300 °C
- 300 °C
- 300 °C
- USP code:
- G14, G15, G16
- G14, G15, G16
- G14, G15, G16
- G14, G15, G16
- G14, G15, G16
- G14, G15, G16
- G14, G15, G16
- G14, G15, G16
- G14, G15, G16
- G14, G15, G16
- G14, G15, G16
- G14, G15, G16
- G14, G15, G16
- G14, G15, G16
- G14, G15, G16
- G14, G15, G16
- G14, G15, G16
- G14, G15, G16
- G14, G15, G16
- G14, G15, G16
- G14, G15, G16
- G14, G15, G16
- G14, G15, G16
- G14, G15, G16